Assay Details
Gene Symbol: LOC101703228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865960.3 | XP_004866017.1 | 7 - 8 | 1795 | 90 | |
XM_013065931.2 | XP_012921385.1 | 7 - 8 | 1796 | 90 | ||
XM_013065933.2 | XP_012921387.1 | 7 - 8 | 1329 | 90 | ||
XM_021245171.1 | XP_021100830.1 | 7 - 8 | 1796 | 90 | ||
XM_021245172.1 | XP_021100831.1 | 7 - 8 | 1797 | 90 | ||
XM_021245173.1 | XP_021100832.1 | 6 - 7 | 1224 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
