Assay Details
Gene Symbol: LOC101703228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865960.3 | XP_004866017.1 | 9 - 10 | 1933 | 70 | |
XM_013065931.2 | XP_012921385.1 | 9 - 10 | 1934 | 70 | ||
XM_013065933.2 | XP_012921387.1 | 9 - 10 | 1467 | 70 | ||
XM_021245171.1 | XP_021100830.1 | 9 - 10 | 1934 | 70 | ||
XM_021245172.1 | XP_021100831.1 | 9 - 10 | 1935 | 70 | ||
XM_021245173.1 | XP_021100832.1 | 8 - 9 | 1362 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
