Assay Details
Gene Symbol: Cfap57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068188.2 | XP_012923642.1 | 5 - 6 | 1106 | 60 | |
XM_021249894.1 | XP_021105553.1 | 5 - 6 | 1106 | 60 | ||
XM_021249895.1 | XP_021105554.1 | 5 - 6 | 1070 | 60 | ||
XM_021249897.1 | XP_021105556.1 | 5 - 6 | 1104 | 60 | ||
XM_021249898.1 | XP_021105557.1 | 5 - 6 | 1123 | 60 | ||
XM_021249899.1 | XP_021105558.1 | 5 - 6 | 1106 | 60 | ||
XM_021249900.1 | XP_021105559.1 | 5 - 6 | 1106 | 60 | ||
XM_021249901.1 | XP_021105560.1 | 5 - 6 | 1106 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
