Assay Details
| Gene Symbol: LOC101702210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250026.1 | XP_021105685.1 | 2 - 3 | 317 | 82 | |
| XM_021250027.1 | XP_021105686.1 | 2 - 3 | 213 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101702210| Gene Symbol: LOC101702210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250026.1 | XP_021105685.1 | 2 - 3 | 317 | 82 | |
| XM_021250027.1 | XP_021105686.1 | 2 - 3 | 213 | 82 | ||