Assay Details
Gene Symbol: LOC101702210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250026.1 | XP_021105685.1 | 4 - 5 | 484 | 64 | |
XM_021250027.1 | XP_021105686.1 | 4 - 5 | 380 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
