Assay Details
| Gene Symbol: LOC101702210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250026.1 | XP_021105685.1 | 5 - 6 | 596 | 67 | |
| XM_021250027.1 | XP_021105686.1 | 5 - 6 | 492 | 67 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  3' Most  Probe spans exons | 
 Back To Top
 Back To Top
 LOC101702210
LOC101702210 Genomic Map
Genomic Map Back To Top
 Back To Top