Assay Details
Gene Symbol: Als2cr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250243.1 | XP_021105902.1 | 3 - 4 | 540 | 62 | |
XM_021250244.1 | XP_021105903.1 | 3 - 4 | 540 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
