Assay Details
Gene Symbol: Znf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840234.3 | XP_004840291.1 | 3 - 4 | 423 | 85 | |
XM_021254529.1 | XP_021110188.1 | 3 - 4 | 370 | 85 | ||
XM_021254530.1 | XP_021110189.1 | 3 - 4 | 290 | 85 | ||
XM_021254531.1 | XP_021110190.1 | 3 - 4 | 366 | 85 | ||
XM_021254532.1 | XP_021110191.1 | 4 - 5 | 346 | 85 | ||
XM_021254533.1 | XP_021110192.1 | 3 - 4 | 349 | 85 | ||
XM_021254534.1 | XP_021110193.1 | 3 - 4 | 248 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
