Assay Details
| Gene Symbol: LOC101700189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853346.3 | XP_004853403.1 | 5 - 6 | 1058 | 95 | |
| XM_004853347.3 | XP_004853404.1 | 5 - 6 | 984 | 95 | ||
More Information
Set Membership: |
|

LOC101700189
Genomic Map