Assay Details
| Gene Symbol: Asphd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856122.3 | XP_004856179.1 | 2 - 3 | 1119 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asphd1| Gene Symbol: Asphd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856122.3 | XP_004856179.1 | 2 - 3 | 1119 | 78 | |
Set Membership: |
|