Assay Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856995.3 | XP_004857052.1 | 4 - 5 | 601 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldoc| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856995.3 | XP_004857052.1 | 4 - 5 | 601 | 62 | |
Set Membership: |
|