Assay Details
| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857078.3 | XP_004857135.1 | 3 - 4 | 363 | 67 | |
| XM_004857079.2 | XP_004857136.1 | 3 - 4 | 241 | 67 | ||
| XM_013076924.2 | XP_012932378.1 | 3 - 4 | 118 | 67 | ||
| XM_021238205.1 | XP_021093864.1 | 3 - 4 | 312 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013076924.2, XM_021238205.1 transcripts.
Set Membership: |
|

Nsrp1
Genomic Map