Assay Details
Gene Symbol: Trim25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859798.2 | XP_004859855.1 | 5 - 6 | 1261 | 62 | |
XM_021239926.1 | XP_021095585.1 | 4 - 5 | 1026 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
