Assay Details
Gene Symbol: Smim22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864824.3 | XP_004864881.1 | 3 - 4 | 315 | 90 | |
XM_004864825.3 | XP_004864882.1 | 3 - 4 | 327 | 90 | ||
XM_004864826.2 | XP_004864883.1 | 3 - 4 | 450 | 90 | ||
XM_021244455.1 | XP_021100114.1 | 3 - 4 | 1427 | 90 | ||
XM_021244456.1 | XP_021100115.1 | 3 - 4 | 323 | 90 | ||
XM_021244457.1 | XP_021100116.1 | 3 - 4 | 293 | 90 | ||
XM_021244458.1 | XP_021100117.1 | 3 - 4 | 419 | 90 | ||
XM_021244459.1 | XP_021100118.1 | 3 - 4 | 833 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
