Assay Details
| Gene Symbol: Pdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872228.2 | XP_004872285.1 | 9 - 10 | 1158 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdk3| Gene Symbol: Pdk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872228.2 | XP_004872285.1 | 9 - 10 | 1158 | 65 | |
Set Membership: |
|