Assay Details
| Gene Symbol: CUNH19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873275.3 | XP_004873332.1 | 2 - 3 | 901 | 94 | |
| XM_004873276.3 | XP_004873333.1 | 2 - 3 | 901 | 94 | ||
| XM_021251075.1 | XP_021106734.1 | 2 - 3 | 901 | 94 | ||
| XM_021251076.1 | XP_021106735.1 | 2 - 3 | 958 | 94 | ||
| XM_021251077.1 | XP_021106736.1 | 2 - 3 | 960 | 94 | ||
| XM_021251078.1 | XP_021106737.1 | 2 - 3 | 901 | 94 | ||

CUNH19orf44
Genomic Map