Assay Details
| Gene Symbol: Znf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068636.2 | XP_012924090.1 | 2 - 3 | 343 | 91 | |
| XM_013068637.2 | XP_012924091.1 | 2 - 3 | 340 | 91 | ||
| XM_013068638.2 | XP_012924092.1 | 2 - 3 | 272 | 91 | ||
More Information
Set Membership: |
|

Znf180
Genomic Map