Assay Details
Gene Symbol: LOC101711115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255598.1 | XP_021111257.1 | 4 - 5 | 824 | 86 | |
XM_021255599.1 | XP_021111258.1 | 4 - 5 | 825 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021255598.1, XM_021255599.1 transcripts.
Set Membership: |
![]() ![]() |
