Assay Details
Gene Symbol: LOC101711115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255598.1 | XP_021111257.1 | 5 - 6 | 936 | 99 | |
XM_021255599.1 | XP_021111258.1 | 5 - 6 | 937 | 99 | ||
XM_021255600.1 | XP_021111259.1 | 6 - 7 | 1031 | 99 | ||
XM_021255601.1 | XP_021111260.1 | 6 - 7 | 1029 | 99 |
