Assay Details
Gene Symbol: LOC101711115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255598.1 | XP_021111257.1 | 6 - 7 | 993 | 82 | |
XM_021255599.1 | XP_021111258.1 | 6 - 7 | 994 | 82 | ||
XM_021255600.1 | XP_021111259.1 | 7 - 8 | 1088 | 82 | ||
XM_021255601.1 | XP_021111260.1 | 7 - 8 | 1086 | 82 |
