Assay Details
Gene Symbol: LOC101711115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255598.1 | XP_021111257.1 | 7 - 8 | 1030 | 106 | |
XM_021255599.1 | XP_021111258.1 | 7 - 8 | 1031 | 106 | ||
XM_021255600.1 | XP_021111259.1 | 8 - 9 | 1125 | 106 | ||
XM_021255601.1 | XP_021111260.1 | 8 - 9 | 1123 | 106 | ||
XM_021255602.1 | XP_021111261.1 | 3 - 4 | 2803 | 106 |

More Information
Set Membership: |
![]() ![]() |
