Assay Details
Gene Symbol: LOC101722877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848053.2 | XP_004848110.1 | 13 - 14 | 2327 | 95 | |
XM_021261056.1 | XP_021116715.1 | 14 - 15 | 2364 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
