Assay Details
Gene Symbol: LOC101722877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848053.2 | XP_004848110.1 | 4 - 5 | 1085 | 108 | |
XM_021261056.1 | XP_021116715.1 | 5 - 6 | 1122 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
