Assay Details
Gene Symbol: Baalc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850243.2 | XP_004850300.1 | 1 - 2 | 181 | 73 | |
XM_004850245.3 | XP_004850302.1 | 1 - 2 | 181 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
