Assay Details
Gene Symbol: Baalc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850243.2 | XP_004850300.1 | 2 - 3 | 337 | 67 | |
XM_021262748.1 | XP_021118407.1 | 2 - 3 | 368 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
