Assay Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851159.3 | XP_004851216.1 | 5 - 6 | 1555 | 69 | |
XM_004851160.3 | XP_004851217.1 | 5 - 6 | 1439 | 69 | ||
XM_013074535.2 | XP_012929989.1 | 5 - 6 | 1442 | 69 | ||
XM_021263569.1 | XP_021119228.1 | 5 - 6 | 1435 | 69 | ||
XM_021263570.1 | XP_021119229.1 | 5 - 6 | 1430 | 69 | ||
XM_021263571.1 | XP_021119230.1 | 5 - 6 | 1286 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
