Assay Details
Gene Symbol: LOC101717478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858887.2 | XP_004858944.1 | 2 - 3 | 1085 | 86 | |
XM_013077470.2 | XP_012932924.1 | 2 - 3 | 1058 | 86 | ||
XM_013077471.2 | XP_012932925.1 | 2 - 3 | 1063 | 86 | ||
XM_021239594.1 | XP_021095253.1 | 2 - 3 | 259 | 86 | ||
XM_021239595.1 | XP_021095254.1 | 2 - 3 | 259 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004858887.2, XM_013077470.2, XM_013077471.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
