Assay Details
Gene Symbol: LOC101717478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858887.2 | XP_004858944.1 | 3 - 4 | 1205 | 76 | |
XM_013077470.2 | XP_012932924.1 | 3 - 4 | 1178 | 76 | ||
XM_013077471.2 | XP_012932925.1 | 3 - 4 | 1183 | 76 | ||
XM_021239594.1 | XP_021095253.1 | 3 - 4 | 379 | 76 |
