Assay Details
Gene Symbol: LOC101717478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858887.2 | XP_004858944.1 | 4 - 5 | 1292 | 107 | |
XM_013077470.2 | XP_012932924.1 | 4 - 5 | 1265 | 107 | ||
XM_013077471.2 | XP_012932925.1 | 4 - 5 | 1270 | 107 | ||
XM_021239594.1 | XP_021095253.1 | 4 - 5 | 466 | 107 | ||
XM_021239595.1 | XP_021095254.1 | 4 - 5 | 463 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
