Assay Details
| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870122.2 | XP_004870179.1 | 5 - 6 | 2004 | 68 | |
| XM_004870123.2 | XP_004870180.1 | 5 - 6 | 2004 | 68 | ||
More Information
Set Membership: |
|

Spag8
Genomic Map