Assay Details
| Gene Symbol: S100a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871437.3 | XP_004871494.1 | 2 - 3 | 390 | 96 | |
| XM_004871439.3 | XP_004871496.1 | 2 - 3 | 313 | 96 | ||
More Information
Set Membership: |
|

S100a16
Genomic Map