Assay Details
Gene Symbol: Tmem132a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874272.3 | XP_004874329.1 | 4 - 5 | 1070 | 65 | |
XM_004874273.3 | XP_004874330.1 | 4 - 5 | 1070 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
