Assay Details
Gene Symbol: LOC101711537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021262276.1 | XP_021117935.1 | 5 - 6 | 460 | 94 | |
XM_021262277.1 | XP_021117936.1 | 3 - 4 | 269 | 94 | ||
XM_021262278.1 | XP_021117937.1 | 3 - 4 | 457 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
