Assay Details
Gene Symbol: LOC101711537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021262276.1 | XP_021117935.1 | 6 - 7 | 618 | 103 | |
XM_021262277.1 | XP_021117936.1 | 4 - 5 | 427 | 103 | ||
XM_021262278.1 | XP_021117937.1 | 4 - 5 | 615 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
