Assay Details
Gene Symbol: Usf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835704.2 | XP_004835761.1 | 3 - 4 | 461 | 71 | |
XM_004835705.2 | XP_004835762.1 | 2 - 3 | 336 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
