Assay Details
Gene Symbol: LOC101716348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844557.3 | XP_004844614.1 | 2 - 3 | 409 | 75 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101716348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844557.3 | XP_004844614.1 | 2 - 3 | 409 | 75 |