Assay Details
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854313.1 | XP_004854370.1 | 3 - 4 | 538 | 79 | |
XM_004854314.2 | XP_004854371.1 | 4 - 5 | 465 | 79 | ||
XM_021266134.1 | XP_021121793.1 | 4 - 5 | 512 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
