Assay Details
Gene Symbol: Rab3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004869252.2 | XP_004869309.1 | 2 - 3 | 385 | 68 | |
XM_004869253.3 | XP_004869310.1 | 2 - 3 | 365 | 68 | ||
XM_021247966.1 | XP_021103625.1 | 2 - 3 | 426 | 68 | ||
XM_021247967.1 | XP_021103626.1 | 2 - 3 | 385 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
