Assay Details
| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874712.3 | XP_004874769.1 | 1 - 2 | 178 | 138 | |
| XM_004874713.3 | XP_004874770.1 | 1 - 2 | 178 | 138 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004874712.3, XM_004874713.3 transcripts.
Set Membership: |
|

Wnt9a
Genomic Map