Assay Details
| Gene Symbol: Wnt9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874712.3 | XP_004874769.1 | 2 - 3 | 439 | 98 | |
| XM_004874713.3 | XP_004874770.1 | 2 - 3 | 439 | 98 | ||
More Information
Set Membership: |
|

Wnt9a
Genomic Map