Assay Details
| Gene Symbol: Znf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069019.2 | XP_012924473.1 | 4 - 5 | 906 | 81 | |
| XM_013069020.2 | XP_012924474.1 | 4 - 5 | 574 | 81 | ||
More Information
Set Membership: |
|

Znf25
Genomic Map