Assay Details
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013074412.2 | XP_012929866.1 | 5 - 6 | 435 | 60 | |
XM_013074413.2 | XP_012929867.1 | 5 - 6 | 456 | 60 | ||
XM_021263262.1 | XP_021118921.1 | 5 - 6 | 489 | 60 | ||
XM_021263263.1 | XP_021118922.1 | 5 - 6 | 501 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
