Assay Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852741.3 | XP_004852798.1 | 2 - 3 | 1273 | 68 | |
| XM_021264798.1 | XP_021120457.1 | 2 - 3 | 1207 | 68 | ||
| XM_021264799.1 | XP_021120458.1 | 2 - 3 | 1206 | 68 | ||
| XM_021264800.1 | XP_021120459.1 | 2 - 3 | 1203 | 68 | ||
| XM_021264802.1 | XP_021120461.1 | 2 - 3 | 1113 | 68 | ||
More Information
Set Membership: |
|

Det1
Genomic Map