Assay Details
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870614.3 | XP_004870671.1 | 9 - 10 | 1652 | 64 | |
XM_013067767.2 | XP_012923221.1 | 9 - 10 | 1905 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
