Assay Details
Gene Symbol: Mcrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873793.3 | XP_004873850.1 | 2 - 3 | 426 | 58 | |
XM_013068911.2 | XP_012924365.1 | 2 - 3 | 426 | 58 | ||
XM_021251445.1 | XP_021107104.1 | 2 - 3 | 550 | 58 |

More Information
Set Membership: |
![]() ![]() |
