Assay Details
Gene Symbol: Mcrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873793.3 | XP_004873850.1 | 4 - 5 | 679 | 68 | |
XM_004873796.3 | XP_004873853.1 | 3 - 4 | 406 | 68 | ||
XM_013068911.2 | XP_012924365.1 | 4 - 5 | 679 | 68 | ||
XM_021251445.1 | XP_021107104.1 | 4 - 5 | 803 | 68 | ||
XM_021251447.1 | XP_021107106.1 | 3 - 4 | 412 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
