Assay Details
Gene Symbol: Mcrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873793.3 | XP_004873850.1 | 5 - 6 | 789 | 93 | |
XM_004873796.3 | XP_004873853.1 | 4 - 5 | 516 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
