Assay Details
| Gene Symbol: LOC106009608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073149.1 | XP_012928603.1 | 2 - 3 | 587 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106009608| Gene Symbol: LOC106009608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073149.1 | XP_012928603.1 | 2 - 3 | 587 | 104 | |
Set Membership: |
|