Assay Details
| Gene Symbol: Rpl26l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239512.1 | XP_021095171.1 | 2 - 3 | 229 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl26l1| Gene Symbol: Rpl26l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239512.1 | XP_021095171.1 | 2 - 3 | 229 | 98 | |
Set Membership: |
|