Assay Details
| Gene Symbol: CUNHXorf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250031.1 | XP_021105690.1 | 2 | 1424 | 80 | |
| XM_021250032.1 | XP_021105691.1 | 2 | 1423 | 80 | ||
| XM_021250033.1 | XP_021105692.1 | 2 | 1377 | 80 | ||
More Information
Set Membership: |
|

CUNHXorf67
Genomic Map